Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01242220_m1
          See other CTCFL GE Assays ›
          Gene Symbol:
          Ctcfl
          Gene Name:
          CCCTC-binding factor (zinc finger protein)-like
          Gene Aliases:
          Boris, OTTMUSG00000016680
          Chromosome Location:
          Chr.2: 173093609 - 173119525 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01242220_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctcfl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081387.2 NP_001074856.1 5 - 6 1306 68
          GenBank mRNA BC151026.1 - 5 - 6 1181 9087939 68
          DQ153171.1 - 5 - 6 1306 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005287.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          regulation of gene expression by genetic imprinting
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          positive regulation of gene expression
          histone methylation
          DNA methylation involved in gamete generation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          regulation of histone H3-K4 methylation
          transcription cis-regulatory region binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          nucleic acid binding
          DNA binding
          protein binding
          histone binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200045066.mm.1 TC0200005287.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045066.mm.1 NM_001081387.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045066.mm.1 NM_001081387.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045071.mm.1 TC0200005287.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045071.mm.1 NM_001081387.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045071.mm.1 NM_001081387.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045068.mm.1 TC0200005287.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045068.mm.1 NM_001081387.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045068.mm.1 NM_001081387.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045067.mm.1 TC0200005287.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045067.mm.1 NM_001081387.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045067.mm.1 NM_001081387.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045072.mm.1 TC0200005287.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045072.mm.1 NM_001081387.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045072.mm.1 NM_001081387.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045070.mm.1 TC0200005287.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045070.mm.1 NM_001081387.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045070.mm.1 NM_001081387.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045069.mm.1 TC0200005287.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045069.mm.1 NM_001081387.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045069.mm.1 NM_001081387.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0