Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01244129_m1
          See other PAPD5 GE Assays ›
          Gene Symbol:
          Papd5
          Gene Name:
          PAP associated domain containing 5
          Gene Aliases:
          5730445M16Rik, 5830428A09
          Chromosome Location:
          Chr.8: 88199213 - 88262997 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01244129_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Papd5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164497.1 NP_001157969.1 12 - 13 2000 64
          NM_001164498.1 NP_001157970.1 12 - 13 1974 64
          NM_001164499.1 NP_001157971.1 11 - 12 1871 64
          GenBank mRNA AK077553.1 - 12 - 13 2002 64
          BC059053.1 - 12 - 13 1972 64
          BC080314.1 - 11 - 12 1575 64
          BC144796.1 - 12 - 13 1958 9053344 64
          BC144797.1 - 11 - 12 1829 9053345 64
          BC145737.1 - 12 - 13 1958 40131040 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001029.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          rRNA processing
          mRNA processing
          cell cycle
          cell division
          histone mRNA catabolic process
          DNA binding
          RNA binding
          DNA-directed DNA polymerase activity
          polynucleotide adenylyltransferase activity
          transferase activity
          nucleotidyltransferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996988 BC059053.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996988 BC080314.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996988 NM_001164497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996988 NM_001164498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996988 NM_001164499.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115268 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145731 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996988 BC059053.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996988 BC080314.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996988 NM_001164497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996988 NM_001164498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996988 NM_001164499.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P204595 AK030850.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P204595 BC059053.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P204595 BC080314.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P204595 NM_001164497.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P204595 NM_001164498.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P239645 BC145737.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P239645 BC144796.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P239645 BC144797.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P239645 AK077553.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P239645 BC059053.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145731 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01789680 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06587079 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01789662 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115268 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06587170 - -
          Whole Mouse Genome G4122A A_52_P204595 AK030850.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204595 BC059053.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204595 BC080314.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204595 NM_001164497.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P204595 NM_001164498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P239645 BC145737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P239645 BC144796.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P239645 BC144797.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P239645 AK077553.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P239645 BC059053.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115268 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07167568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07012420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07012432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07361597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07012428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06942313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07012431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06697066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07012430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26039857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07073682 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10573737 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573746 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573740 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573742 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573743 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573744 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573741 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573739 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573738 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573736 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573735 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573745 - -
          Affymetrix HT MG-430 PM Array Plate 1433738_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10573733 - -
          Mouse Genome 430 2.0 Array 1433738_at - -
          Mouse Genome 430 2.0 Array 1433738_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800007914.mm.1 TC0800001029.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007914.mm.1 NM_001164497.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007914.mm.1 NM_001164497.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007914.mm.1 NM_001164498.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007914.mm.1 NM_001164498.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007914.mm.1 NM_001164499.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007914.mm.1 NM_001164499.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007915.mm.1 TC0800001029.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007915.mm.1 NM_001164497.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007915.mm.1 NM_001164497.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007915.mm.1 NM_001164498.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007915.mm.1 NM_001164498.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007915.mm.1 NM_001164499.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007915.mm.1 NM_001164499.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007917.mm.1 TC0800001029.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007917.mm.1 NM_001164497.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007917.mm.1 NM_001164497.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007917.mm.1 NM_001164498.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007917.mm.1 NM_001164498.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007917.mm.1 NM_001164499.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007917.mm.1 NM_001164499.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007918.mm.1 TC0800001029.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007918.mm.1 NM_001164497.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007918.mm.1 NM_001164497.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007918.mm.1 NM_001164498.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007918.mm.1 NM_001164498.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007918.mm.1 NM_001164499.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007918.mm.1 NM_001164499.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007916.mm.1 TC0800001029.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007916.mm.1 NM_001164497.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007916.mm.1 NM_001164497.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007916.mm.1 NM_001164498.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007916.mm.1 NM_001164498.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007916.mm.1 NM_001164499.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007916.mm.1 NM_001164499.1 Refseq
          Mouse Expression Set 430 1433738_PM_at - -
          Mouse Expression Set 430 1433738_at - -
          Affymetrix HT Mouse Genome 430B Array 1433738_at - -
          Affymetrix HT Mouse Genome 430B Array 1433738_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2644047 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2644047 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0