Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01244534_m1
          See other GM29682 GE Assays ›
          Gene Symbol:
          Gm29682
          Gene Name:
          predicted gene, 29682
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 104088904 - 104110267 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01244534_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm29682
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK033085.1 - 2 - 3 703 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4933400L20 gene 4933400L20Rik -408.5kb Anti-sense Chr. 8 - 103603799 - 103680382 on NCBI Build GRCm38
          Downstream cadherin 5 Cdh5 7B4,
          AA408225,
          Cd144,
          VE-Cad,
          VECD,
          VEcad,
          Vec
          10.9kb Anti-sense Chr. 8 - 104101613 - 104144504 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732583 AF196314.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732583 AK003382.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732583 AK004190.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732583 AK012925.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732583 AK018718.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732583 AK019422.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732583 AK169258.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732583 BC029239.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732583 NM_013897.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 725864 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862653 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 381 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130217 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130217 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126390 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26126666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26126662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26126653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07136484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26126663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26126659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26126658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26126650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07090023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26126660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07136482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26126648 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0