Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000008304.mm.1 |
TC1000001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008304.mm.1 |
NM_001128086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008304.mm.1 |
NM_001128086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008303.mm.1 |
TC1000001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008303.mm.1 |
NM_001128086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008303.mm.1 |
NM_001128086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008306.mm.1 |
TC1000001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008306.mm.1 |
NM_001128086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008306.mm.1 |
NM_001128086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008302.mm.1 |
TC1000001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008302.mm.1 |
NM_001128086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008302.mm.1 |
NM_001128086.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008301.mm.1 |
TC1000001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000008301.mm.1 |
NM_001128086.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000008301.mm.1 |
NM_001128086.2 |
Refseq |