Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01244984_m1
          See other 1700010J16RIK GE Assays ›
          Gene Symbol:
          1700010J16Rik
          Gene Name:
          RIKEN cDNA 1700010J16 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 112726926 - 112785599 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01244984_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700010J16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040579.1 6 - 7 737 74
          GenBank mRNA AK005834.1 - 6 - 7 737 74
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU938461.1 - 295 Chr. - - [-] on Build 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream potassium voltage gated channel,
          Shaw-related subfamily,
          member 2
          Kcnc2 AW047325,
          B230117I07,
          KShIIIA,
          Kv3.2
          -316.4kb Sense Chr. 10 - 112270459 - 112466304 on NCBI Build GRCm38
          Downstream predicted gene 3942 Gm3942 52.1kb Sense Chr. 10 - 112837733 - 112856525 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001313.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 741599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 421625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620764 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 927059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227035 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67174 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549912 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968920 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 70920 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139636 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 57923 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07196116 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07196096 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07196134 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113874 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07196096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08555174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08886733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08826291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08277332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08297234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08579937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08327129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08826292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08219846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08263853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08826306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08300940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27455613 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000009830.mm.1 TC1000001313.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000009830.mm.1 NR_040579.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009830.mm.1 NR_040579.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009831.mm.1 TC1000001313.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000009831.mm.1 NR_040579.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009831.mm.1 NR_040579.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009827.mm.1 TC1000001313.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000009827.mm.1 NR_040579.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000009827.mm.1 NR_040579.1 Refseq
          Murine Genome U74v2 Set 171084_at - -
          Murine Genome U74v2 Set 171084_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 613448 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0