Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01245236_g1
          See other FN3KRP GE Assays ›
          Gene Symbol:
          Fn3krp
          Gene Name:
          fructosamine 3 kinase related protein
          Gene Aliases:
          FN3K-RP
          Chromosome Location:
          Chr.11: 121421373 - 121431288 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01245236_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fn3krp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181420.3 NP_852085.2 5 - 6 684 80
          GenBank mRNA AK078838.1 - 1 - 2 104 80
          AY360466.1 - 5 - 6 642 80
          BC032265.1 - 5 - 6 630 4015688 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002064.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fn3krp

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          phosphorylation
          molecular_function
          phosphofructokinase activity
          phosphorylase kinase regulator activity
          cobinamide kinase activity
          phytol kinase activity
          kinase activity
          phosphatidylinositol phosphate kinase activity
          cyclin-dependent protein serine/threonine kinase regulator activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          phenol kinase activity
          cyclin-dependent protein kinase activating kinase regulator activity
          inositol tetrakisphosphate 2-kinase activity
          heptose 7-phosphate kinase activity
          aminoglycoside phosphotransferase activity
          phosphatidylinositol 3-kinase activity
          eukaryotic elongation factor-2 kinase regulator activity
          eukaryotic elongation factor-2 kinase activator activity
          LPPG:FO 2-phospho-L-lactate transferase activity
          cytidine kinase activity
          glycerate 2-kinase activity
          (S)-lactate 2-kinase activity
          phosphoserine:homoserine phosphotransferase activity
          L-seryl-tRNA(Sec) kinase activity
          phosphocholine transferase activity
          polynucleotide 5'-hydroxyl-kinase activity
          polynucleotide kinase activity
          GTP-dependent polynucleotide kinase activity
          CTP:farnesol kinase activity
          geraniol kinase activity
          geranylgeraniol kinase activity
          CTP:geranylgeraniol kinase activity
          prenol kinase activity
          1-phosphatidylinositol-5-kinase activity
          1-phosphatidylinositol-3-phosphate 4-kinase activity
          phosphatidylinositol-3,4-bisphosphate 5-kinase activity
          phosphatidylinositol bisphosphate kinase activity
          inositol-3,4,6-trisphosphate 1-kinase activity
          inositol 5-diphosphate pentakisphosphate 5-kinase activity
          inositol diphosphate tetrakisphosphate kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28190418 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28190421 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418147 AK078838.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418147 NM_181420.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418147 AK078838.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P418147 NM_181420.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297931 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352878 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54666 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177917 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418147 AK078838.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418147 NM_181420.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418147 AK078838.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P418147 NM_181420.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28190405 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02448217 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177917 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07519076 - -
          Whole Mouse Genome G4122A A_51_P418147 AK078838.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418147 NM_181420.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418147 AK078838.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418147 NM_181420.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P835149 AK034934.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101317 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177917 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09435054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09624377 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09624368 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09624369 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09624372 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09435050 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28190421 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28190418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28190375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09706519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09363483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09140822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09529678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09086222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09113272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09140821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09074928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09529666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09529675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09074926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09529677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09324620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09435143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09435142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09183048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28190472 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10383558 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383562 - -
          Affymetrix HT MG-430 PM Array Plate 1443050_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10383556 - -
          Affymetrix Mouse Clariom S Array PSR1100019941.mm.1 TC1100002064.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100019941.mm.1 NM_181420.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100019941.mm.1 NM_181420.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 861004 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3060128 - -
          Illumina mouse-6 v1.0 expression beadchip 3060128 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2773586 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2773586 - -
          mouse-6 v1.1 (Illumina) scl40614.6.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 3060128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40614.6.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060128 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2913980 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2773585 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2913980 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40614.6.1_28-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix