Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01245681_m1
          See other CAMTA2 GE Assays ›
          Gene Symbol:
          Camta2
          Gene Name:
          calmodulin binding transcription activator 2
          Gene Aliases:
          Kiaa0909-hp, mKIAA0909
          Chromosome Location:
          Chr.11: 70669463 - 70688108 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01245681_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Camta2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190376.1 NP_001177305.1 12 - 13 2306 104
          NM_001190378.1 NP_001177307.1 12 - 13 2306 104
          NM_001190379.1 NP_001177308.1 11 - 12 2234 104
          NM_178116.4 NP_835217.1 12 - 13 2321 104
          GenBank mRNA AK039670.1 - 12 - 13 2133 104
          AK122400.1 - 10 - 11 1962 104
          AK150186.1 - 12 - 13 2159 104
          AK169118.1 - 11 - 12 2073 104
          BC023962.1 - 7 - 8 2120 104
          BC027385.1 - 4 - 5 1112 104
          BC049133.1 - 12 - 13 2150 6827800 104
          BC056395.1 - 12 - 13 2135 6852349 104
          BC059065.1 - 11 - 12 2101 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003110.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Camta2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          cardiac muscle hypertrophy in response to stress
          positive regulation of transcription by RNA polymerase II
          DNA binding
          chromatin binding
          transcription factor binding
          histone deacetylase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100028734.mm.1 TC1100003110.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028734.mm.1 NM_001190376.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028734.mm.1 NM_001190376.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028734.mm.1 NM_001190378.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028734.mm.1 NM_001190378.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028734.mm.1 NM_001190379.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028734.mm.1 NM_001190379.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028734.mm.1 NM_178116.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028734.mm.1 NM_178116.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028735.mm.1 TC1100003110.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028735.mm.1 NM_001190376.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028735.mm.1 NM_001190376.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028735.mm.1 NM_001190378.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028735.mm.1 NM_001190378.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028735.mm.1 NM_001190379.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028735.mm.1 NM_001190379.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028735.mm.1 NM_178116.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028735.mm.1 NM_178116.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix