Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01246298_g1
          See other TRPV1 GE Assays ›
          Gene Symbol:
          Trpv1
          Gene Name:
          transient receptor potential cation channel, subfamily V, member 1
          Gene Aliases:
          OTRPC1, TRPV1alpha, TRPV1beta, VR-1, Vr1
          Chromosome Location:
          Chr.11: 73231024 - 73261322 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01246298_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpv1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001445.2 NP_001001445.1 4 - 5 857 68
          GenBank mRNA AB180097.1 - 3 - 4 605 68
          AJ620495.1 - 3 - 4 645 68
          AK051315.1 - 4 - 5 715 68
          AY445519.1 - 3 - 4 605 68
          AY452083.1 - 3 - 4 638 68
          AY452084.1 - 3 - 4 638 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001019.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trpv1

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          fever generation
          microglial cell activation
          diet induced thermogenesis
          peptide secretion
          lipid metabolic process
          transport
          ion transport
          cation transport
          calcium ion transport
          signal transduction
          positive regulation of cytosolic calcium ion concentration
          response to pH
          response to heat
          response to organonitrogen compound
          glutamate secretion
          urinary bladder smooth muscle contraction
          sensory perception of pain
          ion transmembrane transport
          catagen
          positive regulation of apoptotic process
          positive regulation of nitric oxide biosynthetic process
          response to pain
          behavioral response to pain
          sensory perception of mechanical stimulus
          thermoception
          detection of chemical stimulus involved in sensory perception of pain
          transmembrane transport
          smooth muscle contraction involved in micturition
          positive regulation of gastric acid secretion
          calcium ion transmembrane transport
          cellular response to alkaloid
          cellular response to ATP
          cellular response to cytokine stimulus
          negative regulation of establishment of blood-brain barrier
          cation transmembrane transport
          nucleotide binding
          transmembrane signaling receptor activity
          ion channel activity
          excitatory extracellular ligand-gated ion channel activity
          cation channel activity
          calcium channel activity
          protein binding
          calmodulin binding
          ATP binding
          cation transmembrane transporter activity
          ligand-gated ion channel activity
          calcium-release channel activity
          chloride channel regulator activity
          identical protein binding
          phosphoprotein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100009261.mm.1 TC1100001019.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009261.mm.1 NM_001001445.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0