Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01246836_m1
          See other 6820408C15RIK GE Assays ›
          Gene Symbol:
          6820408C15Rik
          Gene Name:
          RIKEN cDNA 6820408C15 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 152415587 - 152444336 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01246836_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6820408C15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289738.1 NP_001276667.1 11 - 12 1424 66
          NM_177656.4 NP_808324.1 11 - 12 1421 66
          NR_110367.1 11 - 12 1462 66
          NR_110368.1 10 - 11 1362 66
          GenBank mRNA AK078482.1 - 11 - 12 1421 66
          AW048149.1 - 1 - 2 66
          BC137826.1 - 10 - 11 1301 8860845 66
          BC144943.1 - 10 - 11 1339 66
          BC144944.1 - 10 - 11 1301 9053492 66
          BC144945.1 - 9 - 10 1235 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002282.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 6820408C15Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344942 BC144944.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344942 BC137826.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344942 NM_177656.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344942 AK078482.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344942 BC144944.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344942 BC137826.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344942 NM_177656.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344942 AK078482.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156244 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898713 BC116754.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898713 BC116756.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898713 NM_147108.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 61335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 398019 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300823 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 758003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10924 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159729 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168226 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344942 BC144944.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344942 BC137826.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344942 NM_177656.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344942 AK078482.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344942 BC144944.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344942 BC137826.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344942 NM_177656.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344942 AK078482.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21749244 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04741841 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159729 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168226 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00528508 - -
          Whole Mouse Genome G4122A A_51_P344942 BC144944.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344942 BC137826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344942 NM_177656.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344942 AK078482.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344942 BC144944.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344942 BC137826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344942 NM_177656.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P344942 AK078482.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111742 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106603 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168226 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159729 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01318128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21749313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01318127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01930282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01371061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01655458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02196289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01930272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02267720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02282069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21749291 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10477114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477110 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477108 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477111 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477109 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477119 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477112 - -
          Affymetrix HT MG-430 PM Array Plate 1441306_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10477107 - -
          Mouse Genome 430 2.0 Array 1441306_at - -
          Mouse Genome 430 2.0 Array 1442366_PM_at - -
          Mouse Genome 430 2.0 Array 1442366_at - -
          Mouse Genome 430 2.0 Array 1441306_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 228778_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 228778_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200017890.mm.1 TC0200002282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017890.mm.1 NM_001289738.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017890.mm.1 NM_177656.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017890.mm.1 NR_110367.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017890.mm.1 NR_110368.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017889.mm.1 TC0200002282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017889.mm.1 NM_001289738.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017889.mm.1 NM_177656.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017889.mm.1 NR_110367.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017889.mm.1 NR_110368.1 Refseq
          Mouse Expression Set 430 1441306_at - -
          Mouse Expression Set 430 1441306_PM_at - -
          Murine Genome U74v2 Set 129855_PM_g_at - -
          Murine Genome U74v2 Set 129855_g_at - -
          Murine Genome U74v2 Set 129854_PM_at - -
          Murine Genome U74v2 Set 129854_at - -
          Affymetrix HT Mouse Genome 430B Array 1441306_at - -
          Affymetrix HT Mouse Genome 430B Array 1441306_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4730112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004730112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245721 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245721 - -
          mouse-6 v1.1 (Illumina) scl20121.12.1_122-S - -
          Illumina Sentrix mouse-6.v1 bead array 4730112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20121.12.1_122-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4730112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245721 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245721 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fcd8d4bc9-cwdc5:80/100.66.130.62:80.
          git-commit: a06ae585138c3108a562191b4d7dc526e77a8a21
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0