Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01249101_g1
          See other CES1H GE Assays ›
          Gene Symbol:
          Ces1h
          Gene Name:
          carboxylesterase 1H
          Gene Aliases:
          2310039D24Rik
          Chromosome Location:
          Chr.8: 93351842 - 93386990 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01249101_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ces1h
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK009689.1 - 5 - 6 422 109

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342200 AK009689.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P342200 AK009689.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162905 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342200 AK009689.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P342200 AK009689.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162905 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06599956 - -
          Whole Mouse Genome G4122A A_51_P342200 AK009689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P342200 AK009689.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125039 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07159653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07197117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06894874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06924829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07197124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26071318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06924834 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432261_PM_at - -
          Mouse Genome 430 2.0 Array 1432261_at - -
          Mouse Genome 430 2.0 Array 1432261_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75704_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75704_at - -
          Mouse Expression Set 430 1432261_at - -
          Mouse Expression Set 430 1432261_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432261_at - -
          Affymetrix HT Mouse Genome 430B Array 1432261_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2230575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241347 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241347 - -
          mouse-6 v1.1 (Illumina) scl34486.9.1_200-S - -
          Illumina Sentrix mouse-6.v1 bead array 2230575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34486.9.1_200-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230575 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241347 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix