Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01249473_m1
          See other 4930422I22RIK GE Assays ›
          Gene Symbol:
          4930422I22Rik
          Gene Name:
          RIKEN cDNA 4930422I22 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01249473_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930422I22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015180.1 - 2 - 3 773 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream tetraspanin 8 Tspan8 C76990,
          E330007O21Rik,
          Tm4sf3
          Anti-sense Chr. 10 - 115816817 - 115849893 on NCBI Build GRCm38
          Downstream protein tyrosine phosphatase,
          receptor type,
          R
          Ptprr Gmcp1,
          PTP-SL,
          PTPBR7,
          RPTPRR,
          mPTP213
          168.2kb Anti-sense Chr. 10 - 116018200 - 116274932 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2401194 AK015180.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 702357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334282 AK037774.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334282 AK048683.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334282 AK051503.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334282 XM_001474947.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610147 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965596 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 523117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33008 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 62939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 113219 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104198 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02226074 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07202852 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104198 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128762 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08911496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08218021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08845595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08469130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08304485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08701560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08218020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27472141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27471711 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432049_PM_at - -
          Mouse Genome 430 2.0 Array 1432049_at - -
          Mouse Genome 430 2.0 Array 1432049_PM_at - -
          Mouse Expression Set 430 1432049_at - -
          Mouse Expression Set 430 1432049_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432049_at - -
          Affymetrix HT Mouse Genome 430B Array 1432049_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0