Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800020840.mm.1 |
TC0800002722.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020840.mm.1 |
NM_011787.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020840.mm.1 |
NM_011787.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020842.mm.1 |
TC0800002722.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020842.mm.1 |
NM_011787.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020842.mm.1 |
NM_011787.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020845.mm.1 |
TC0800002722.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020845.mm.1 |
NM_011787.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020845.mm.1 |
NM_011787.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020841.mm.1 |
TC0800002722.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020841.mm.1 |
NM_011787.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020841.mm.1 |
NM_011787.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020844.mm.1 |
TC0800002722.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020844.mm.1 |
NM_011787.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020844.mm.1 |
NM_011787.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020839.mm.1 |
TC0800002722.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020839.mm.1 |
NM_011787.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020839.mm.1 |
NM_011787.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020846.mm.1 |
TC0800002722.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020846.mm.1 |
NM_011787.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020846.mm.1 |
NM_011787.2 |
Refseq |