Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200044351.mm.1 |
TC0200005181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200044351.mm.1 |
NM_001033196.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044351.mm.1 |
NM_001033196.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044354.mm.1 |
TC0200005181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200044354.mm.1 |
NM_001033196.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044354.mm.1 |
NM_001033196.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044349.mm.1 |
TC0200005181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200044349.mm.1 |
NM_001033196.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044349.mm.1 |
NM_001033196.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044353.mm.1 |
TC0200005181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200044353.mm.1 |
NM_001033196.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044353.mm.1 |
NM_001033196.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044352.mm.1 |
TC0200005181.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200044352.mm.1 |
NM_001033196.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200044352.mm.1 |
NM_001033196.2 |
Refseq |