Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01250354_m1
          See other RPAIN GE Assays ›
          Gene Symbol:
          Rpain
          Gene Name:
          RPA interacting protein
          Gene Aliases:
          2400006N03Rik, Rip
          Chromosome Location:
          Chr.11: 70970200 - 70977933 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01250354_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpain
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252415.1 NP_001239344.1 6 - 7 837 62
          NM_027186.2 NP_081462.1 6 - 7 931 62
          GenBank mRNA AK010289.1 - 6 - 7 660 62
          BC047996.1 - 6 - 7 918 6509910 62
          BY576068.1 - 1 - 2 141 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000973.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpain

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA-dependent DNA replication
          DNA repair
          DNA recombination
          protein import into nucleus
          response to UV
          protein-containing complex binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143261 BC047996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2143261 NM_027186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P253697 BC047996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P253697 NM_027186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P253697 AK010289.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P253697 BC047996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P253697 NM_027186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P253697 AK010289.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325933 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 497891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590661 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 1943 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P253697 BC047996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P253697 NM_027186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P253697 AK010289.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P253697 BC047996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P253697 NM_027186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P253697 AK010289.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143261 BC047996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2143261 NM_027186.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27903624 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02358261 - -
          Whole Mouse Genome G4122A A_51_P253697 BC047996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P253697 NM_027186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P253697 AK010289.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P253697 BC047996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P253697 NM_027186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P253697 AK010289.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09651035 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09651034 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09651040 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09651037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27903602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09651040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09113699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09651037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09086853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09595119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09113706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09738043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09676765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27903640 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10378021 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378017 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378015 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378023 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378014 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378020 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378016 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378019 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378022 - -
          Affymetrix HT MG-430 PM Array Plate 1441814_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1457621_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428828_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10378013 - -
          Mouse Genome 430 2.0 Array 1441814_PM_s_at - -
          Mouse Genome 430 2.0 Array 1441814_s_at - -
          Mouse Genome 430 2.0 Array 1457621_at - -
          Mouse Genome 430 2.0 Array 1428828_at - -
          Mouse Genome 430 2.0 Array 1428828_PM_at - -
          Mouse Genome 430 2.0 Array 1457621_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69723_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69723_at - -
          Affymetrix Mouse Clariom S Array PSR1100008744.mm.1 TC1100000973.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008744.mm.1 NM_001252413.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008744.mm.1 NM_001252413.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008744.mm.1 NM_001252414.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008744.mm.1 NM_001252414.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008744.mm.1 NM_001252415.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008744.mm.1 NM_001252415.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008744.mm.1 NM_027186.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008744.mm.1 NM_027186.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008739.mm.1 TC1100000973.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008739.mm.1 NM_001252415.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008739.mm.1 NM_001252415.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008739.mm.1 NM_027186.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008739.mm.1 NM_027186.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008741.mm.1 TC1100000973.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008741.mm.1 NM_001252413.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008741.mm.1 NM_001252413.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008741.mm.1 NM_001252414.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008741.mm.1 NM_001252414.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008741.mm.1 NM_001252415.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008741.mm.1 NM_001252415.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008741.mm.1 NM_027186.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008741.mm.1 NM_027186.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008740.mm.1 TC1100000973.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008740.mm.1 NM_001252413.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008740.mm.1 NM_001252413.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008740.mm.1 NM_001252414.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008740.mm.1 NM_001252414.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008740.mm.1 NM_001252415.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008740.mm.1 NM_001252415.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008740.mm.1 NM_027186.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008740.mm.1 NM_027186.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008745.mm.1 TC1100000973.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008745.mm.1 NM_001252413.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008745.mm.1 NM_001252413.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008745.mm.1 NM_001252414.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008745.mm.1 NM_001252414.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008745.mm.1 NM_001252415.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008745.mm.1 NM_001252415.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008745.mm.1 NM_027186.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008745.mm.1 NM_027186.2 Refseq
          Mouse Expression Set 430 1428828_PM_at - -
          Mouse Expression Set 430 1441814_PM_s_at - -
          Mouse Expression Set 430 1457621_PM_at - -
          Mouse Expression Set 430 1441814_s_at - -
          Mouse Expression Set 430 1457621_at - -
          Mouse Expression Set 430 1428828_at - -
          Affymetrix HT Mouse Genome 430B Array 1428828_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441814_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1457621_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457621_at - -
          Affymetrix HT Mouse Genome 430B Array 1428828_at - -
          Affymetrix HT Mouse Genome 430B Array 1441814_s_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100380136 - -
          Illumina mouse-6 v1.0 expression beadchip 100380136 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2762050 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247991 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223766 - -
          mouse-6 v1.1 (Illumina) scl0001528.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 100380136 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100380136 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2968807 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247991 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2968807 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0