Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045531.1 | NP_001038996.1 | 4 - 5 | 336 | 65 | |
GenBank mRNA | BC147586.1 | - | 4 - 5 | 318 | 9056576 | 65 |
BC147607.1 | - | 4 - 5 | 318 | 9056597 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |