Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01251832_m1
          See other SPATA22 GE Assays ›
          Gene Symbol:
          Spata22
          Gene Name:
          spermatogenesis associated 22
          Gene Aliases:
          Gm882
          Chromosome Location:
          Chr.11: 73326537 - 73346434 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01251832_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spata22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001045531.1 NP_001038996.1 5 - 6 429 145
          GenBank mRNA BC147586.1 - 5 - 6 411 9056576 145
          BC147607.1 - 5 - 6 411 9056597 145

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001021.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic DNA repair synthesis
          homologous chromosome pairing at meiosis
          gamete generation
          fertilization
          meiotic cell cycle
          regulation of meiotic cell cycle
          reproductive system development
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27916627 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022123 NM_001045531.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 100205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 658858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172116 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022123 NM_001045531.1 mouse_transcriptome
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27916627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27916778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09637440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08983015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09596590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09438996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09342283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09628159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09200800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09420971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09745075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09012363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27916637 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10378392 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378388 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378386 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378391 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378390 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378387 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378393 - -
          Affymetrix Mouse Gene 1.1 ST Array 10378385 - -
          Affymetrix Mouse Clariom S Array PSR1100009308.mm.1 TC1100001021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009308.mm.1 NM_001045531.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009308.mm.1 NM_001045531.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009311.mm.1 TC1100001021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009311.mm.1 NM_001045531.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009311.mm.1 NM_001045531.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009312.mm.1 TC1100001021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009312.mm.1 NM_001045531.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009312.mm.1 NM_001045531.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009313.mm.1 TC1100001021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009313.mm.1 NM_001045531.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009313.mm.1 NM_001045531.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009309.mm.1 TC1100001021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009309.mm.1 NM_001045531.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009309.mm.1 NM_001045531.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009310.mm.1 TC1100001021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009310.mm.1 NM_001045531.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009310.mm.1 NM_001045531.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 521118 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239455 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2957488 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2957488 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0