Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01254236_m1
          See other TDRD1 GE Assays ›
          Gene Symbol:
          Tdrd1
          Gene Name:
          tudor domain containing 1
          Gene Aliases:
          MTR-1
          Chromosome Location:
          Chr.19: 56826171 - 56870012 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01254236_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tdrd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001002238.2 NP_001002238.1 6 - 7 765 122
          NM_001002240.2 NP_001002240.1 6 - 7 738 122
          NM_001002241.2 NP_001002241.1 5 - 6 1038 122
          NM_031387.3 NP_113564.2 6 - 7 898 122
          GenBank mRNA AB067571.1 - 5 - 6 719 122
          AB183526.1 - 6 - 7 765 122
          AB183527.1 - 6 - 7 738 122
          AB183528.1 - 5 - 6 1038 122
          AB183529.1 - 5 - 6 602 122
          AF285591.1 - 2 - 3 92 122
          AK029806.1 - 6 - 7 899 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000809.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AB183529.1, AF285591.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tdrd1

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          germ cell development
          spermatogenesis
          cell differentiation
          gene silencing by RNA
          piRNA metabolic process
          DNA methylation involved in gamete generation
          meiotic cell cycle
          protein binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900007094.mm.1 TC1900000809.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900007094.mm.1 NM_001002238.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007094.mm.1 NM_001002238.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007094.mm.1 NM_001002240.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007094.mm.1 NM_001002240.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007094.mm.1 NM_001002241.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007094.mm.1 NM_001002241.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007094.mm.1 NM_031387.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007094.mm.1 NM_031387.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007093.mm.1 TC1900000809.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900007093.mm.1 NM_001002238.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007093.mm.1 NM_001002238.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007093.mm.1 NM_001002240.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007093.mm.1 NM_001002240.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007093.mm.1 NM_001002241.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007093.mm.1 NM_001002241.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007093.mm.1 NM_031387.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900007093.mm.1 NM_031387.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0