Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285839.1 | NP_001272768.1 | 1 - 2 | 235 | 91 | |
NR_104360.1 | 1 - 2 | 235 | 91 | |||
NR_104361.1 | 1 - 2 | 235 | 91 | |||
GenBank mRNA | AK011265.1 | - | 1 - 2 | 183 | 91 | |
AK045669.1 | - | 1 - 2 | 197 | 91 | ||
AK137332.1 | - | 1 - 2 | 186 | 91 | ||
AK137402.1 | - | 1 - 2 | 94 | 91 | ||
BC030671.1 | - | 1 - 2 | 205 | 91 | ||
BC058172.1 | - | 1 - 2 | 198 | 5012054 | 91 | |
BY780319.1 | - | 1 - 2 | 236 | 91 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |