Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01254990_m1
          See other ABI2 GE Assays ›
          Gene Symbol:
          Abi2
          Gene Name:
          abl-interactor 2
          Gene Aliases:
          8430425M24Rik, AI839867, C130078H13
          Chromosome Location:
          Chr.1: 60408988 - 60481945 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01254990_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abi2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001198570.1 NP_001185499.1 2 - 3 464 130
          NM_001198571.1 NP_001185500.1 2 - 3 464 130
          NM_198127.2 NP_937760.1 2 - 3 464 130
          GenBank mRNA BC056345.1 - 2 - 3 285 5705048 130
          BC079545.1 - 2 - 3 349 6851145 130
          BC079646.1 - 2 - 3 388 6850720 130

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000471.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abi2

          Gene Ontology Categories:

          Function(s) Process(es)

          movement of cell or subcellular component
          learning or memory
          actin polymerization or depolymerization
          dendrite development
          cell migration
          Rac protein signal transduction
          peptidyl-tyrosine phosphorylation
          camera-type eye development
          positive regulation of Arp2/3 complex-mediated actin nucleation
          SH3 domain binding
          small GTPase binding
          ubiquitin protein ligase binding
          protein-containing complex binding
          proline-rich region binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100003931.mm.1 TC0100000471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100003931.mm.1 NM_001198570.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003931.mm.1 NM_001198570.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003931.mm.1 NM_001198571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003931.mm.1 NM_001198571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003931.mm.1 NM_198127.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003931.mm.1 NM_198127.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003933.mm.1 TC0100000471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100003933.mm.1 NM_001198570.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003933.mm.1 NM_001198570.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003933.mm.1 NM_001198571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003933.mm.1 NM_001198571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003933.mm.1 NM_198127.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003933.mm.1 NM_198127.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003929.mm.1 TC0100000471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100003929.mm.1 NM_001198570.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003929.mm.1 NM_001198570.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003929.mm.1 NM_001198571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003929.mm.1 NM_001198571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003929.mm.1 NM_198127.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003929.mm.1 NM_198127.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003930.mm.1 TC0100000471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100003930.mm.1 NM_001198570.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003930.mm.1 NM_001198570.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003930.mm.1 NM_001198571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003930.mm.1 NM_001198571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003930.mm.1 NM_198127.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003930.mm.1 NM_198127.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003932.mm.1 TC0100000471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100003932.mm.1 NM_001198570.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003932.mm.1 NM_001198570.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003932.mm.1 NM_001198571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003932.mm.1 NM_001198571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003932.mm.1 NM_198127.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003932.mm.1 NM_198127.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003928.mm.1 TC0100000471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100003928.mm.1 NM_001198570.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003928.mm.1 NM_001198570.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003928.mm.1 NM_001198571.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003928.mm.1 NM_001198571.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003928.mm.1 NM_198127.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100003928.mm.1 NM_198127.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0