Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100019425.mm.1 |
TC0100002385.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019425.mm.1 |
NM_172513.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019425.mm.1 |
NM_172513.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019427.mm.1 |
TC0100002385.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019427.mm.1 |
NM_172513.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019427.mm.1 |
NM_172513.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019428.mm.1 |
TC0100002385.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019428.mm.1 |
NM_172513.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019428.mm.1 |
NM_172513.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019424.mm.1 |
TC0100002385.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019424.mm.1 |
NM_172513.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019424.mm.1 |
NM_172513.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019426.mm.1 |
TC0100002385.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100019426.mm.1 |
NM_172513.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100019426.mm.1 |
NM_172513.3 |
Refseq |