Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01256912_g1
          See other APH1A GE Assays ›
          Gene Symbol:
          Aph1a
          Gene Name:
          anterior pharynx defective 1a homolog (C. elegans)
          Gene Aliases:
          6530402N02Rik, APH-1a
          Chromosome Location:
          Chr.3: 95893907 - 95898592 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01256912_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aph1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146104.3 NP_666216.1 1 - 2 361 89
          NM_146134.2 NP_666246.1 1 - 2 361 89
          GenBank mRNA AK148400.1 - 1 - 2 314 89
          AK152334.1 - 1 - 2 304 89
          AX775953.1 - 1 - 2 229 89
          BC012406.1 - 1 - 2 286 3483214 89
          BC024111.1 - 1 - 2 227 5098973 89
          BC057865.1 - 1 - 2 304 5320175 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000914.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aph1a

          Gene Ontology Categories:

          Function(s) Process(es)

          metanephros development
          proteolysis
          membrane protein ectodomain proteolysis
          Notch signaling pathway
          Notch receptor processing
          protein processing
          membrane protein intracellular domain proteolysis
          amyloid precursor protein catabolic process
          positive regulation of catalytic activity
          endopeptidase activity
          protein binding
          peptidase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300007273.mm.1 TC0300000914.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007273.mm.1 NM_146104.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007273.mm.1 NM_146134.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007276.mm.1 TC0300000914.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007276.mm.1 NM_146104.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007276.mm.1 NM_146134.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007270.mm.1 TC0300000914.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007270.mm.1 NM_146104.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007270.mm.1 NM_146134.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007274.mm.1 TC0300000914.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007274.mm.1 NM_146104.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007274.mm.1 NM_146134.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 903223 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0