Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01257388_m1
          See other RNF165 GE Assays ›
          Gene Symbol:
          Rnf165
          Gene Name:
          ring finger protein 165
          Gene Aliases:
          2900024M11Rik, AI427432, Akd2, Ark2c, G630064H08Rik, Gm96
          Chromosome Location:
          Chr.18: 77456110 - 77566378 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01257388_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf165
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164504.1 NP_001157976.1 3 - 4 524 57
          GenBank mRNA BY727947.1 - 2 - 3 438 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001625.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          axonogenesis
          motor neuron axon guidance
          multicellular organism aging
          protein catabolic process
          positive regulation of BMP signaling pathway
          forelimb morphogenesis
          limb development
          innervation
          muscle structure development
          protein binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P220262 BC066016.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P220262 NM_001164504.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P220262 AK013591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P220262 BC066016.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P220262 NM_001164504.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P220262 AK013591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117664 AK133955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117664 NM_001164504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117664 AK133955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117664 NM_001164504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P220262 BC066016.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P220262 NM_001164504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P220262 AK013591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P220262 BC066016.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P220262 NM_001164504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P220262 AK013591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236629 BC066016.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236629 AK036050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P236629 NM_001164504.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220262 BC066016.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220262 NM_001164504.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220262 AK013591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220262 BC066016.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220262 NM_001164504.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220262 AK013591.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P13893370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13996412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31848005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14045377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13818769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31848016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31848020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13810316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31848014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31848000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31848011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31848008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13976733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31848003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13893371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31848019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13818771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13818757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13818767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13810314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31848001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13996404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31848002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31847973 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1436114_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1458303_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438829_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10459827 - -
          Mouse Genome 430 2.0 Array 1447404_at - -
          Mouse Genome 430 2.0 Array 1447404_PM_at - -
          Mouse Genome 430 2.0 Array 1458303_PM_at - -
          Mouse Genome 430 2.0 Array 1458303_at - -
          Mouse Genome 430 2.0 Array 1438829_at - -
          Mouse Genome 430 2.0 Array 1438829_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225743_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225743_at - -
          Affymetrix Mouse Clariom S Array PSR1800011703.mm.1 TC1800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011703.mm.1 NM_001164504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011703.mm.1 NM_001164504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011702.mm.1 TC1800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011702.mm.1 NM_001164504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011702.mm.1 NM_001164504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011710.mm.1 TC1800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011710.mm.1 NM_001164504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011710.mm.1 NM_001164504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011704.mm.1 TC1800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011704.mm.1 NM_001164504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011704.mm.1 NM_001164504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011711.mm.1 TC1800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011711.mm.1 NM_001164504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011711.mm.1 NM_001164504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011701.mm.1 TC1800001625.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011701.mm.1 NM_001164504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011701.mm.1 NM_001164504.1 Refseq
          Affymetrix HT Mouse Genome 430B Array 1447404_at - -
          Affymetrix HT Mouse Genome 430B Array 1458303_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458303_at - -
          Affymetrix HT Mouse Genome 430B Array 1447404_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436114_at - -
          Affymetrix HT Mouse Genome 430B Array 1436114_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438829_at - -
          Affymetrix HT Mouse Genome 430B Array 1438829_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 781531 - -
          ABI Mouse Genome Survey Microarray 807416 - -
          ABI Mouse Genome Survey Microarray 865481 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259244 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2590554 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2726781 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0