Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afg3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027130.1 | NP_081406.1 | 13 - 14 | 1795 | 67 | |
GenBank mRNA | AK080118.1 | - | 8 - 9 | 2830 | 67 | |
AK149970.1 | - | 13 - 14 | 1767 | 67 | ||
AK151207.1 | - | 13 - 14 | 1816 | 67 | ||
AK152115.1 | - | 13 - 14 | 1826 | 67 | ||
AK152727.1 | - | 13 - 14 | 1826 | 67 | ||
AK164537.1 | - | 13 - 14 | 1836 | 67 | ||
AK168668.1 | - | 13 - 14 | 1855 | 67 | ||
BC021380.1 | - | 13 - 14 | 1792 | 67 | ||
BC036999.1 | - | 13 - 14 | 1839 | 5042649 | 67 | |
BC043056.1 | - | 13 - 14 | 1795 | 6419521 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() |