Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146057.1 | NP_001139529.1 | 3 - 4 | 521 | 67 | |
NM_001146058.1 | NP_001139530.1 | 3 - 4 | 383 | 67 | ||
NM_133348.2 | NP_579926.2 | 3 - 4 | 510 | 67 | ||
GenBank mRNA | AB049821.1 | - | 3 - 4 | 476 | 67 | |
AB088411.1 | - | 3 - 4 | 449 | 67 | ||
AB088412.1 | - | 3 - 4 | 417 | 67 | ||
AB207243.1 | - | 3 - 4 | 476 | 67 | ||
AK152277.1 | - | 3 - 4 | 391 | 67 | ||
BC013507.1 | - | 3 - 4 | 378 | 4224774 | 67 | |
CA463703.1 | - | 513 | 67 | |||
CX225643.1 | - | 4 - 5 | 414 | 68 | ||
CX235974.1 | - | 3 - 4 | 402 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |