Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264642_m1
          See other GM17751 GE Assays ›
          Gene Symbol:
          Gm17751
          Gene Name:
          predicted gene, 17751
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 76489133 - 76502028 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264642_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm17751
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038013.1 4 - 5 309 73
          GenBank mRNA AK054265.1 - 4 - 5 309 73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CO807055.1 - 4 - 5 283 Chr. 1 - 76489159 - 76500833 [+] on Build GRCm38 73
          GenBankmRNA BB608842.2 - 4 - 5 309 Chr. 1 - 76489133 - 76500702 [+] on Build GRCm38 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier family 4 (anion exchanger),
          member 3
          Slc4a3 A930038D23Rik,
          Ae3
          -938.1kb Sense Chr. 1 - 75546041 - 75559431 on NCBI Build GRCm38
          Downstream microRNA 6343 Mir6343 Gm27304,
          mmu-mir-6343
          8.7kb Sense Chr. 1 - 76509560 - 76509643 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2368607 NR_038012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2368607 NR_038013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2368607 AK054265.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932431 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473077 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 405398 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134967 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2368607 NR_038012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2368607 NR_038013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2368607 AK054265.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04152997 - -
          Whole Mouse Genome G4122A A_52_P699405 NR_038012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P699405 NR_038013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P699405 AK054265.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00646856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00525835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00554456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00456172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00128908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00083718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00732801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00710503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00554458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00160774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00083720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00985186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00533173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00083715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00808340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00554455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00858537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00732794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00808338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00821700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00554454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00821701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01131210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00985185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00710507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00879391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20359972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00462204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20360057 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 422756 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0