Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264771_m1
          See other 8430422H06RIK GE Assays ›
          Gene Symbol:
          8430422H06Rik
          Gene Name:
          RIKEN cDNA 8430422H06 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 14232306 - 14278542 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264771_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 8430422H06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045373.1 3 - 4 493 71
          GenBank mRNA AK018433.1 - 3 - 4 496 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger protein 521 Zfp521 B930086A16Rik,
          Evi3,
          Znf521
          -288.6kb Anti-sense Chr. 18 - 13687013 - 13972787 on NCBI Build GRCm38
          Downstream synovial sarcoma translocation,
          Chromosome 18
          Ss18 D130059H17,
          Ssxt,
          Syt
          347.1kb Anti-sense Chr. 18 - 14625629 - 14683002 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000118.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2245532 AK018433.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 373226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30899 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129216 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445058 AK010792.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445058 AK012729.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445058 AK019289.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445058 AK020514.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445058 AK020515.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445058 BC012031.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445058 BC016449.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445058 NM_019403.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 146134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52296 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08918858 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08918876 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08918832 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126015 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08918832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13932804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13957060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14118743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13919115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13682168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13996558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13596344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13913114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14086036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13973831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14086017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14086032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13933735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13957063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13996560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13957058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31511464 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432165_PM_at - -
          Mouse Genome 430 2.0 Array 1432165_PM_at - -
          Mouse Genome 430 2.0 Array 1432165_at - -
          Affymetrix Mouse Clariom S Array PSR1800000878.mm.1 TC1800000118.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000878.mm.1 NR_045373.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000878.mm.1 NR_045373.1 Refseq
          Mouse Expression Set 430 1432165_PM_at - -
          Mouse Expression Set 430 1432165_at - -
          Affymetrix HT Mouse Genome 430B Array 1432165_at - -
          Affymetrix HT Mouse Genome 430B Array 1432165_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 650617 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105890500 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0