Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264834_m1
          See other 4930546C10RIK GE Assays ›
          Gene Symbol:
          4930546C10Rik
          Gene Name:
          RIKEN cDNA 4930546C10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 68889487 - 68951528 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264834_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930546C10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038051.1 6 - 7 2060 75
          GenBank mRNA AK019744.1 - 6 - 7 2061 75

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 6356 Mir6356 Gm27461,
          mmu-mir-6356
          -150.0kb Anti-sense Chr. 18 - 68739400 - 68739501 on NCBI Build GRCm38
          Downstream microRNA 145b Mir145b Gm27644,
          mmu-mir-145b
          131.8kb Sense Chr. 18 - 69022199 - 69022273 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001526.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121607 AK019744.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121607 NR_038051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121607 AK019744.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P121607 NR_038051.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148314 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134911 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121607 AK019744.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121607 NR_038051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121607 AK019744.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P121607 NR_038051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121607 AK019744.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121607 NR_038051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121607 AK019744.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P121607 NR_038051.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13707875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14109339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14022812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14004285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13970350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13723153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13970358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13697382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13608102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13710057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14038145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13940427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14109335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13864085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14118462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13711767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14118458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13901822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14026191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14025870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13848580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14100051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14026096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13794785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13710313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14114361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13832123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14010876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14026192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13970362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13824670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31799913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31800380 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431846_PM_at - -
          Mouse Genome 430 2.0 Array 1431846_at - -
          Mouse Genome 430 2.0 Array 1431846_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800011186.mm.1 TC1800001526.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011186.mm.1 NR_038051.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011186.mm.1 NR_038051.1 Refseq
          Mouse Expression Set 430 1431846_PM_at - -
          Mouse Expression Set 430 1431846_at - -
          Affymetrix HT Mouse Genome 430B Array 1431846_at - -
          Affymetrix HT Mouse Genome 430B Array 1431846_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 646022 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0