Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900015566.mm.1 |
TC1900001762.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015566.mm.1 |
NM_009970.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015566.mm.1 |
NM_009970.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015560.mm.1 |
TC1900001762.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015560.mm.1 |
NM_009970.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015560.mm.1 |
NM_009970.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015561.mm.1 |
TC1900001762.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015561.mm.1 |
NM_009970.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015561.mm.1 |
NM_009970.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015563.mm.1 |
TC1900001762.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015563.mm.1 |
NM_009970.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015563.mm.1 |
NM_009970.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015564.mm.1 |
TC1900001762.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015564.mm.1 |
NM_009970.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015564.mm.1 |
NM_009970.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015562.mm.1 |
TC1900001762.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015562.mm.1 |
NM_009970.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015562.mm.1 |
NM_009970.2 |
Refseq |