Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01267315_m1
          See other ZYG11A GE Assays ›
          Gene Symbol:
          Zyg11a
          Gene Name:
          zyg-11 family member A, cell cycle regulator
          Gene Aliases:
          BC022150
          Chromosome Location:
          Chr.4: 108180245 - 108218160 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01267315_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zyg11a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167936.1 NP_001161408.1 12 - 13 1997 99
          GenBank mRNA BC022150.1 - 10 - 11 1545 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003236.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          regulation of ubiquitin-protein transferase activity
          molecular_function
          ubiquitin-protein transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026609 BC022150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026609 NM_001167936.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127553 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026609 BC022150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026609 NM_001167936.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05353231 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05353246 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127553 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03804802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03548580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04029217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03861528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03804808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23194995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03905467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23194990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04049092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03686462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23194993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23194985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23194992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23194999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10514975 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514982 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514980 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514976 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514978 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514974 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514983 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514984 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514981 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514979 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514977 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514973 - -
          Affymetrix HT MG-430 PM Array Plate 1427647_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10514971 - -
          Mouse Genome 430 2.0 Array 1427647_PM_at - -
          Mouse Genome 430 2.0 Array 1427647_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230590_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230590_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400025760.mm.1 TC0400003236.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400025760.mm.1 NM_001167936.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400025761.mm.1 TC0400003236.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400025761.mm.1 NM_001167936.2 Refseq
          Mouse Expression Set 430 1427647_at - -
          Mouse Expression Set 430 1427647_PM_at - -
          Mouse Genome 430A 2.0 Array 1427647_PM_at - -
          Mouse Genome 430A 2.0 Array 1427647_at - -
          Affymetrix HT Mouse Genome 430A Array 1427647_at - -
          Affymetrix HT Mouse Genome 430A Array 1427647_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 829346 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0