Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028132.3 | NP_082408.3 | 10 - 11 | 1717 | 99 | |
GenBank mRNA | AK011485.1 | - | 10 - 11 | 1642 | 99 | |
AK147982.1 | - | 10 - 11 | 1689 | 99 | ||
AK152678.1 | - | 10 - 11 | 1717 | 99 | ||
AK152923.1 | - | 10 - 11 | 1717 | 99 | ||
BC005617.1 | - | 2 - 3 | 180 | 99 | ||
BC008527.1 | - | 10 - 11 | 1661 | 3495694 | 99 | |
BC036290.1 | - | 10 - 11 | 1905 | 99 | ||
BC055713.1 | - | 10 - 11 | 1668 | 99 | ||
BC080801.1 | - | 10 - 11 | 1688 | 99 | ||
BC086490.1 | - | 10 - 11 | 1622 | 30435602 | 99 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |