Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01268229_m1
          See other PGM2 GE Assays ›
          Gene Symbol:
          Pgm2
          Gene Name:
          phosphoglucomutase 2
          Gene Aliases:
          2610020G18Rik, AA407108, AI098105, Pgm-2, Pgm1
          Chromosome Location:
          Chr.4: 99929451 - 99987294 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01268229_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pgm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028132.3 NP_082408.3 3 - 4 674 76
          GenBank mRNA AK011485.1 - 3 - 4 599 76
          AK147982.1 - 3 - 4 646 76
          AK152678.1 - 3 - 4 674 76
          AK152923.1 - 3 - 4 674 76
          AK212160.1 - 2 - 3 289 76
          BC008527.1 - 3 - 4 618 3495694 76
          BC036290.1 - 3 - 4 862 76
          BC055713.1 - 3 - 4 625 76
          BC080801.1 - 3 - 4 645 76
          BC086490.1 - 3 - 4 579 30435602 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001010.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pgm2

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          glycogen biosynthetic process
          glucose metabolic process
          galactose catabolic process
          organic substance metabolic process
          magnesium ion binding
          phosphoglucomutase activity
          isomerase activity
          intramolecular transferase activity, phosphotransferases
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400006832.mm.1 TC0400001010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006832.mm.1 NM_028132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006832.mm.1 NM_028132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006834.mm.1 TC0400001010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006834.mm.1 NM_028132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006834.mm.1 NM_028132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006836.mm.1 TC0400001010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006836.mm.1 NM_028132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006836.mm.1 NM_028132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006830.mm.1 TC0400001010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006830.mm.1 NM_028132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006830.mm.1 NM_028132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006837.mm.1 TC0400001010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006837.mm.1 NM_028132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006837.mm.1 NM_028132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006833.mm.1 TC0400001010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006833.mm.1 NM_028132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006833.mm.1 NM_028132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006835.mm.1 TC0400001010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006835.mm.1 NM_028132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006835.mm.1 NM_028132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006828.mm.1 TC0400001010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006828.mm.1 NM_028132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006828.mm.1 NM_028132.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006829.mm.1 TC0400001010.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400006829.mm.1 NM_028132.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400006829.mm.1 NM_028132.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 645843 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0