Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138655.1 | NP_619596.1 | 14 - 15 | 2137 | 116 | |
GenBank mRNA | AF417581.1 | - | 14 - 15 | 2137 | 116 | |
AY263156.1 | - | 3 - 4 | 304 | 116 |
Set Membership: |
![]() ![]() ![]() ![]() |