Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128080.2 | NP_001121552.1 | 2 - 3 | 241 | 65 | |
NM_001128081.1 | NP_001121553.1 | 1 - 2 | 356 | 65 | ||
NM_001128082.1 | NP_001121554.1 | 1 - 2 | 356 | 65 | ||
NM_020286.3 | NP_064682.1 | 1 - 2 | 356 | 65 | ||
GenBank mRNA | AF175771.1 | - | 1 - 2 | 205 | 65 | |
AF291425.1 | - | 1 - 2 | 324 | 65 | ||
AJ279791.1 | - | 1 - 2 | 316 | 65 | ||
AJ279792.1 | - | 1 - 2 | 316 | 65 | ||
AJ279793.1 | - | 1 - 2 | 316 | 65 | ||
AJ279794.1 | - | 1 - 2 | 316 | 65 | ||
AJ279795.1 | - | 1 - 2 | 316 | 65 | ||
AK013248.1 | - | 1 - 2 | 132 | 65 | ||
AK041748.1 | - | 2 - 3 | 227 | 65 | ||
AK154694.1 | - | 1 - 2 | 377 | 65 | ||
AK171199.1 | - | 1 - 2 | 355 | 65 | ||
AY039000.1 | - | 2 - 3 | 249 | 65 | ||
BC051204.1 | - | 1 - 2 | 346 | 65 | ||
BC055858.1 | - | 1 - 2 | 330 | 4913671 | 65 | |
BY208601.1 | - | 1 - 2 | 374 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |