Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01268887_m1
          See other TSPAN32 GE Assays ›
          Gene Symbol:
          Tspan32
          Gene Name:
          tetraspanin 32
          Gene Aliases:
          AW208513, Art-1, BB235973, D7Wsu37e, Phemx, Tssc6
          Chromosome Location:
          Chr.7: 143004902 - 143019486 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01268887_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tspan32
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001128080.2 NP_001121552.1 3 - 4 340 76
          NM_001128081.1 NP_001121553.1 2 - 3 455 76
          NM_001128082.1 NP_001121554.1 2 - 3 455 76
          NM_020286.3 NP_064682.1 2 - 3 455 76
          GenBank mRNA AF175771.1 - 2 - 3 304 76
          AF291425.1 - 2 - 3 423 76
          AJ279791.1 - 2 - 3 415 76
          AJ279792.1 - 2 - 3 415 76
          AJ279793.1 - 2 - 3 415 76
          AJ279794.1 - 2 - 3 415 76
          AJ279795.1 - 2 - 3 415 76
          AK013248.1 - 2 - 3 231 76
          AK154694.1 - 2 - 3 476 76
          AK171199.1 - 2 - 3 455 76
          AY039000.1 - 3 - 4 348 76
          BC051204.1 - 2 - 3 445 76
          BC055858.1 - 2 - 3 429 4913671 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002087.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tspan32

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          integrin-mediated signaling pathway
          blood coagulation
          hemostasis
          negative regulation of cell population proliferation
          negative regulation of myeloid dendritic cell activation
          defense response to protozoan
          regulation of defense response to virus
          platelet aggregation
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700018803.mm.1 TC0700002087.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700018803.mm.1 NM_001128080.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018803.mm.1 NM_001128080.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018803.mm.1 NM_001128081.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018803.mm.1 NM_001128081.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018803.mm.1 NM_001128082.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018803.mm.1 NM_001128082.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018803.mm.1 NM_020286.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700018803.mm.1 NM_020286.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0