Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01274207_m1
          See other XNTRPC GE Assays ›
          Gene Symbol:
          Xntrpc Trpc2
          Gene Name:
          Xndc1-transient receptor potential cation channel, subfamily C, member 2 readthrough
          transient receptor potential cation channel, subfamily C, member 2
          Gene Aliases:
          Xndr-trpc2
          3010009O07Rik, AI115608, TRPC2a, TRPC2b, Trrp2, mTrp2, smTRPC2, trp2
          Chromosome Location:
          Chr.7: 102065491 - 102096864 on Build GRCm38
          Chromosome Location:
          Chr.7: 102083116 - 102096864 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm01274207_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xntrpc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011644.3 NP_035774.2 12 - 13 2175 71
          NR_104582.1 12 - 13 2738 71
          GenBank mRNA AK040538.1 - 12 - 13 2516 71
          Gene Symbol: Trpc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109897.2 NP_001103367.1 3 - 4 1011 71
          GenBank mRNA AF230803.1 - 3 - 4 1011 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001446.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Xntrpc
          Click here to find siRNAs for Gene Trpc2

          Gene Ontology Categories:

          Function(s) Process(es)

          single strand break repair
          inter-male aggressive behavior
          territorial aggressive behavior
          transport
          ion transport
          calcium ion transport
          manganese ion transport
          positive regulation of cytosolic calcium ion concentration
          acrosome reaction
          female courtship behavior
          regulation of gene expression
          response to pheromone
          mating behavior, sex discrimination
          transmembrane transport
          calcium ion transmembrane transport
          negative regulation of male mating behavior
          damaged DNA binding
          ion channel activity
          calcium channel activity
          protein binding
          calmodulin binding
          store-operated calcium channel activity
          diacylglycerol binding
          inositol 1,4,5 trisphosphate binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700012519.mm.1 TC0700001446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012519.mm.1 NM_001109897.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012519.mm.1 NM_001286689.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012519.mm.1 NM_001286690.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012523.mm.1 TC0700001446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012523.mm.1 NM_001109897.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012523.mm.1 NM_011644.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012523.mm.1 NR_104582.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012524.mm.1 TC0700001446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012524.mm.1 NM_001109897.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012524.mm.1 NM_011644.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012524.mm.1 NR_104582.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012516.mm.1 TC0700001446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012516.mm.1 NM_001286689.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012516.mm.1 NM_001286690.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012516.mm.1 NM_011644.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix