Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01274290_g1
          See other ILK GE Assays ›
          Gene Symbol:
          Ilk
          Gene Name:
          integrin linked kinase
          Gene Aliases:
          AA511515, ESTM24
          Chromosome Location:
          Chr.7: 105736590 - 105742925 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01274290_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ilk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161724.1 NP_001155196.1 1 - 2 128 79
          GenBank mRNA AK002344.1 - 1 - 2 46 79
          AK011766.1 - 1 - 2 66 79
          AK154207.1 - 1 - 2 49 79
          CA752166.1 - 1 - 2 128 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001557.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001161724.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          branching involved in ureteric bud morphogenesis
          positive regulation of cell-matrix adhesion
          protein phosphorylation
          negative regulation of protein kinase activity
          integrin-mediated signaling pathway
          cell aging
          positive regulation of cell population proliferation
          regulation of signal transduction
          negative regulation of cardiac muscle cell apoptotic process
          fibroblast migration
          Schwann cell development
          negative regulation of smooth muscle cell migration
          phosphorylation
          peptidyl-serine phosphorylation
          nerve development
          myelination in peripheral nervous system
          cell projection organization
          positive regulation of cell migration
          positive regulation of BMP signaling pathway
          myelin assembly
          regulation of actin cytoskeleton organization
          substrate adhesion-dependent cell spreading
          positive regulation of phosphorylation
          negative regulation of apoptotic process
          positive regulation of MAP kinase activity
          positive regulation of MAPK cascade
          protein kinase B signaling
          negative regulation of neuron apoptotic process
          establishment or maintenance of epithelial cell apical/basal polarity
          positive regulation of myoblast differentiation
          positive regulation of osteoblast differentiation
          positive regulation of axon extension
          positive regulation of transcription, DNA-templated
          negative regulation of smooth muscle cell proliferation
          neuron projection morphogenesis
          positive regulation of axonogenesis
          positive regulation of dendrite morphogenesis
          protein heterooligomerization
          positive regulation of protein kinase B signaling
          platelet aggregation
          positive regulation of canonical Wnt signaling pathway
          supramolecular fiber organization
          negative regulation of neural precursor cell proliferation
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          integrin binding
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          SH3 domain binding
          protein kinase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700013082.mm.1 TC0700001557.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013082.mm.1 NM_001161724.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013082.mm.1 NM_001161724.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013081.mm.1 TC0700001557.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013081.mm.1 NM_001161724.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013081.mm.1 NM_001161724.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c8579b687-54cxl:80/100.66.135.11:80.
          git-commit: 8e87205ac1279514f9c7a0d34bcd4a43061862b7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0