Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700036462.mm.1 |
TC0700004340.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036462.mm.1 |
NM_024414.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036462.mm.1 |
NM_024414.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036460.mm.1 |
TC0700004340.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036460.mm.1 |
NM_024414.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036460.mm.1 |
NM_024414.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036461.mm.1 |
TC0700004340.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036461.mm.1 |
NM_024414.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036461.mm.1 |
NM_024414.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036463.mm.1 |
TC0700004340.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036463.mm.1 |
NM_024414.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036463.mm.1 |
NM_024414.2 |
Refseq |