Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01275959_g1
          See other RBBP9 GE Assays ›
          Gene Symbol:
          Rbbp9
          Gene Name:
          retinoblastoma binding protein 9
          Gene Aliases:
          Bog
          Chromosome Location:
          Chr.2: 144542265 - 144550859 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01275959_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbbp9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015754.2 NP_056569.2 4 - 5 412 90
          GenBank mRNA AF039563.1 - 4 - 5 331 90
          AK017606.1 - 4 - 5 353 90
          AK076944.1 - 4 - 5 407 90
          BC011107.1 - 4 - 5 350 3982875 90
          BC147814.1 - 4 - 5 405 9087531 90
          BC147815.1 - 4 - 5 405 9087532 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004763.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbbp9

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell population proliferation
          molecular_function
          GPI-anchor transamidase activity
          hydrolase activity
          coenzyme F390-A hydrolase activity
          coenzyme F390-G hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707836 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707834 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707833 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707835 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707838 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707837 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006615 AK076944.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006615 BC011107.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006615 BC050201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006615 NM_015754.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006615 AK003857.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178592 BC147814.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178592 BC147815.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178592 AK076944.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178592 BC011107.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178592 BC050201.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 279042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 590919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720440 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884273 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 154773 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125899 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178592 BC147814.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178592 BC147815.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178592 AK076944.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178592 BC011107.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178592 BC050201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006615 AK076944.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006615 BC011107.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006615 BC050201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006615 NM_015754.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006615 AK003857.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P299125 AF039563.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21707807 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125899 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04724457 - -
          Whole Mouse Genome G4122A A_51_P299125 AF039563.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P483721 AK076944.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P483721 BC011107.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P483721 BC050201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P483721 NM_015754.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P483721 AK003857.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122611 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04724457 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125899 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02022892 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01396179 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01396174 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01396170 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02022897 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02022896 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707838 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707836 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707834 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707833 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707835 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21707837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21707842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02058698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02058680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01885810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01423092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01702222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01907420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01702224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01724124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02058696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01907415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01724120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01885807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02058687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02058681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02058704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21707809 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10488296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488298 - -
          Affymetrix HT MG-430 PM Array Plate 1416174_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10488291 - -
          Mouse Genome 430 2.0 Array 1416174_PM_at - -
          Mouse Genome 430 2.0 Array 1416174_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26450_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26450_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200040809.mm.1 TC0200004763.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200040809.mm.1 NM_015754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040809.mm.1 NM_015754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040810.mm.1 TC0200004763.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200040810.mm.1 NM_015754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040810.mm.1 NM_015754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040811.mm.1 TC0200004763.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200040811.mm.1 NM_015754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040811.mm.1 NM_015754.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040812.mm.1 TC0200004763.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200040812.mm.1 NM_015754.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040812.mm.1 NM_015754.2 Refseq
          Mouse Expression Set 430 1416174_at - -
          Mouse Expression Set 430 1416174_PM_at - -
          Murine Genome U74v2 Set 104618_PM_at - -
          Murine Genome U74v2 Set 93222_at - -
          Murine Genome U74v2 Set 162230_PM_r_at - -
          Murine Genome U74v2 Set 93222_PM_at - -
          Murine Genome U74v2 Set 162230_r_at - -
          Murine Genome U74v2 Set 104618_at - -
          Mouse Genome 430A 2.0 Array 1416174_PM_at - -
          Mouse Genome 430A 2.0 Array 1416174_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 625143 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103990735 - -
          Illumina mouse-6 v1.0 expression beadchip 2480139 - -
          Illumina mouse-6 v1.0 expression beadchip 103990735 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002480139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2635623 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214213 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2635623 - -
          mouse-6 v1.1 (Illumina) scl0026450.2_100-S - -
          mouse-6 v1.1 (Illumina) scl18572.5_283-S - -
          Illumina Sentrix mouse-6.v1 bead array 2480139 - -
          Illumina Sentrix mouse-6.v1 bead array 103990735 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0026450.2_100-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2480139 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103990735 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2890496 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214213 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222004 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2890496 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0