Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177469.3 | NP_803420.2 | 1 - 2 | 361 | 81 | |
GenBank mRNA | AK082207.1 | - | 1 - 2 | 240 | 81 | |
BE651866.1 | - | 1 - 2 | 327 | 81 | ||
BG083237.2 | - | 1 - 2 | 361 | 81 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |