Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01276555_m1
          See other ATF2 GE Assays ›
          Gene Symbol:
          Atf2
          Gene Name:
          activating transcription factor 2
          Gene Aliases:
          Atf-2, CRE-BP, Creb2, D130078H02Rik, D18875, Tg(Gzma-Klra1)7Wum, mXBP
          Chromosome Location:
          Chr.2: 73816509 - 73892639 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01276555_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025093.2 NP_001020264.1 14 - 15 1666 83
          NM_001284369.1 NP_001271298.1 12 - 13 1506 83
          NM_001284372.1 NP_001271301.1 12 - 13 1372 83
          NM_001284373.1 NP_001271302.1 11 - 12 1309 83
          NM_001284374.1 NP_001271303.1 11 - 12 1391 83
          NM_001284376.1 NP_001271305.1 14 - 15 1548 83
          NM_009715.3 NP_033845.1 13 - 14 1569 83
          GenBank mRNA AF483482.1 - 10 - 11 1243 83
          AF483483.1 - 10 - 11 1243 83
          AK046184.1 - 3 - 4 427 83
          AK051591.1 - 13 - 14 1559 83
          AK139515.1 - 12 - 13 1422 83
          AK144977.1 - 12 - 13 1346 83
          AK145887.1 - 12 - 13 1276 83
          AY902311.1 - 9 - 10 1129 83
          BC042210.1 - 11 - 12 1342 4018323 83
          BC079883.1 - 13 - 14 1491 6839572 83
          BC082596.1 - 11 - 12 1255 30649207 83
          BC085137.1 - 11 - 12 1334 83
          M31629.1 - 6 - 7 721 83
          M77167.1 - 8 - 9 1015 83
          S76657.1 - 1334 83
          S76659.1 - 1040 83
          U46026.1 - 3 - 4 511 83

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atf2

          Gene Ontology Categories:

          Function(s) Process(es)

          outflow tract morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          response to osmotic stress
          cellular response to DNA damage stimulus
          histone acetylation
          mitotic intra-S DNA damage checkpoint signaling
          positive regulation of transforming growth factor beta2 production
          fat cell differentiation
          positive regulation of transcription by RNA polymerase II
          positive regulation of DNA-binding transcription factor activity
          adipose tissue development
          positive regulation of mitochondrial membrane permeability involved in apoptotic process
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          nucleic acid binding
          DNA binding
          chromatin binding
          DNA-binding transcription factor activity
          histone acetyltransferase activity
          protein binding
          cAMP response element binding protein binding
          transferase activity
          protein kinase binding
          cAMP response element binding
          sequence-specific DNA binding
          metal ion binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0