Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01277547_g1
          See other 1700120G07RIK GE Assays ›
          Gene Symbol:
          1700120G07Rik
          Gene Name:
          RIKEN cDNA 1700120G07 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 135191765 - 135196002 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01277547_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700120G07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046050.1 3 - 4 407 90
          GenBank mRNA AK007216.1 - 3 - 4 408 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY707307.1 - 3 - 4 411 Chr. 7 - 135191765 - 135196002 [-] on Build GRCm38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream dedicator of cytokinesis 1 Dock1 9130006G06Rik,
          AI854900,
          D630004B07Rik
          -18.1kb Anti-sense Chr. 7 - 134670608 - 135173647 on NCBI Build GRCm38
          Downstream neuropeptide S Nps ENSMUSG00000073804 75.6kb Anti-sense Chr. 7 - 135268619 - 135272942 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004447.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264030 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133119 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133119 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06393966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05941142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06443353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06466973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06577060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05956701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06512912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06580818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06609070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06393956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06577056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06105191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06393955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06577059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06362290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25551167 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700037357.mm.1 TC0700004447.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037357.mm.1 NR_046050.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037357.mm.1 NR_046050.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037360.mm.1 TC0700004447.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037360.mm.1 NR_046050.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037360.mm.1 NR_046050.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037361.mm.1 TC0700004447.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037361.mm.1 NR_046050.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037361.mm.1 NR_046050.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037358.mm.1 TC0700004447.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037358.mm.1 NR_046050.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037358.mm.1 NR_046050.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037363.mm.1 TC0700004447.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037363.mm.1 NR_046050.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037363.mm.1 NR_046050.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 305023 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101400592 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix