Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175113.3 | NP_780322.2 | 1 - 2 | 247 | 77 | |
GenBank mRNA | AK014359.1 | - | 1 - 2 | 244 | 77 | |
AK017672.1 | - | 1 - 2 | 189 | 77 | ||
AK028755.1 | - | 1 - 2 | 247 | 77 | ||
AK129300.1 | - | 1 - 2 | 214 | 77 | ||
AK164396.1 | - | 1 - 2 | 180 | 77 | ||
AK165976.1 | - | 1 - 2 | 199 | 77 | ||
AK167222.1 | - | 1 - 2 | 213 | 77 | ||
BC027565.1 | - | 1 - 2 | 153 | 77 | ||
BC052648.1 | - | 1 - 2 | 165 | 30030801 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |