Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01283155_m1
          See other PGRMC2 GE Assays ›
          Gene Symbol:
          Pgrmc2
          Gene Name:
          progesterone receptor membrane component 2
          Gene Aliases:
          4631434O19Rik, 5730409G06Rik, DG6, PMBP
          Chromosome Location:
          Chr.3: 41066326 - 41083046 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01283155_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pgrmc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027558.1 NP_081834.1 2 - 3 596 82
          GenBank mRNA AK014543.1 - 2 - 3 495 82
          AK184727.1 - 1 - 2 146 82
          AX775753.1 - 2 - 3 552 82
          BC044759.2 - 2 - 3 537 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001909.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          steroid binding
          protein binding
          lipid binding
          heme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413916 AX775753.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413916 BC044759.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413916 AK140064.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413916 NM_027558.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413916 AK014543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413916 AX775753.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413916 BC044759.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413916 AK140064.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413916 NM_027558.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413916 AK014543.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6753 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779298 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471272 AK035542.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471272 AK083256.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471272 AK150971.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471272 AK151267.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471272 AK162081.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471272 BC054112.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471272 BC080290.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471272 NM_001003948.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 372986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110487 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37809 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134189 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154913 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413916 AX775753.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413916 BC044759.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413916 AK140064.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413916 NM_027558.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413916 AK014543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413916 AX775753.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413916 BC044759.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413916 AK140064.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413916 NM_027558.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413916 AK014543.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154913 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134189 - -
          Whole Mouse Genome G4122A A_51_P413916 AX775753.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413916 BC044759.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413916 AK140064.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413916 NM_027558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413916 AK014543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413916 AX775753.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413916 BC044759.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413916 AK140064.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413916 NM_027558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413916 AK014543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335326 AX775753.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335326 BC044759.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335326 AK140064.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335326 AK165560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335326 NM_027558.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134189 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02813134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02799737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03118862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02936185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02667618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03176626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02545389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02545395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02372345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02799734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02813137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22082989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22083045 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10497967 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497966 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497965 - -
          Affymetrix HT MG-430 PM Array Plate 1452882_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428714_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10497964 - -
          Mouse Genome 430 2.0 Array 1452882_PM_at - -
          Mouse Genome 430 2.0 Array 1428714_PM_at - -
          Mouse Genome 430 2.0 Array 1452882_at - -
          Mouse Genome 430 2.0 Array 1428714_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70804_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70804_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300014643.mm.1 TC0300001909.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300014643.mm.1 NM_027558.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014643.mm.1 NM_027558.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014642.mm.1 TC0300001909.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300014642.mm.1 NM_027558.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014642.mm.1 NM_027558.1 Refseq
          Mouse Expression Set 430 1428714_PM_at - -
          Mouse Expression Set 430 1428714_at - -
          Mouse Expression Set 430 1452882_PM_at - -
          Mouse Expression Set 430 1452882_at - -
          Murine Genome U74v2 Set 94801_PM_at - -
          Murine Genome U74v2 Set 161447_PM_f_at - -
          Murine Genome U74v2 Set 94801_at - -
          Murine Genome U74v2 Set 161447_f_at - -
          Mouse Genome 430A 2.0 Array 1428714_PM_at - -
          Mouse Genome 430A 2.0 Array 1428714_at - -
          Mouse Genome 430A 2.0 Array 1452882_at - -
          Mouse Genome 430A 2.0 Array 1452882_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452882_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428714_at - -
          Affymetrix HT Mouse Genome 430A Array 1428714_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452882_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 375231 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 103520673 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220641 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2715141 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2524166 - -
          Illumina Sentrix mouse-6.v1 bead array 103520673 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103520673 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220641 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix