Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166653.1 | NP_001160125.1 | 7 - 8 | 1101 | 159 | |
NM_001166654.1 | NP_001160126.1 | 7 - 8 | 1191 | 159 | ||
NM_001166655.1 | NP_001160127.1 | 7 - 8 | 1194 | 159 | ||
NM_001166656.1 | NP_001160128.1 | 7 - 8 | 1101 | 159 | ||
NM_153785.4 | NP_722480.3 | 7 - 8 | 1191 | 159 | ||
GenBank mRNA | AK045036.1 | - | 6 - 7 | 1550 | 159 | |
AK045356.1 | - | 7 - 8 | 1091 | 159 | ||
AK046394.1 | - | 7 - 8 | 1192 | 159 | ||
AK050990.1 | - | 7 - 8 | 1191 | 159 | ||
AK078804.1 | - | 7 - 8 | 1083 | 159 | ||
AK090045.1 | - | 7 - 8 | 977 | 159 | ||
AK166428.1 | - | 7 - 8 | 1033 | 159 |
Set Membership: |
![]() ![]() ![]() ![]() |