Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
TC1100000585.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_001166653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_001166653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_001166654.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_001166654.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_001166655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_001166655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_001166656.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_001166656.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_001166657.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_001166657.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_153785.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004550.mm.1 |
NM_153785.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
TC1100000585.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_001166653.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_001166653.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_001166654.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_001166654.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_001166655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_001166655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_001166656.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_001166656.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_001166657.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_001166657.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_153785.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004551.mm.1 |
NM_153785.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004553.mm.1 |
TC1100000585.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004553.mm.1 |
NM_001166655.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004553.mm.1 |
NM_001166655.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004553.mm.1 |
NM_001166657.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004553.mm.1 |
NM_001166657.1 |
Refseq |