Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF128214.1 | - | 3 - 4 | 391 | 96 | |
AF128215.1 | - | 3 - 4 | 391 | 96 | ||
AK162981.1 | - | 3 - 4 | 415 | 96 | ||
BC103608.1 | - | 3 - 4 | 397 | 40046200 | 96 | |
BC103609.1 | - | 3 - 4 | 397 | 40046202 | 96 | |
BC103610.1 | - | 3 - 4 | 397 | 40046204 | 96 | |
BC103614.1 | - | 3 - 4 | 397 | 40046206 | 96 | |
M86671.1 | - | 3 - 4 | 392 | 96 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |