Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01289049_g1
          See other RAB29 GE Assays ›
          Gene Symbol:
          Rab29
          Gene Name:
          RAB29, member RAS oncogene family
          Gene Aliases:
          Rab7l1
          Chromosome Location:
          Chr.1: 131867210 - 131872887 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01289049_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab29
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144875.2 NP_659124.2 5 - 6 763 91
          GenBank mRNA AB232625.1 - 4 - 5 496 91
          AK079752.1 - 4 - 5 617 91
          BC016133.1 - 5 - 6 748 4485029 91
          BC029056.1 - 4 - 5 537 4947517 91
          CK796080.1 - 1 - 2 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001177.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab29

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          Golgi organization
          small GTPase mediated signal transduction
          protein transport
          cell differentiation
          melanosome organization
          Rab protein signal transduction
          retrograde transport, plasma membrane to Golgi
          positive regulation of intracellular protein transport
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20618102 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20618101 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20618104 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20618103 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P112164 CK796080.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P112164 AK185980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P112164 BC016133.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P112164 BC029056.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P112164 NM_144875.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077079 AK079752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077079 AK185980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077079 BC016133.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077079 BC029056.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077079 NM_144875.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729899 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141994 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077079 AK079752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077079 AK185980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077079 BC016133.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077079 BC029056.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077079 NM_144875.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P112164 CK796080.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P112164 AK185980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P112164 BC016133.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P112164 BC029056.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P112164 NM_144875.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377004 CK796080.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377004 BC016133.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377004 BC029056.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377004 NM_144875.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20618102 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141994 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00186894 - -
          Whole Mouse Genome G4122A A_51_P377004 CK796080.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377004 BC016133.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377004 BC029056.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377004 NM_144875.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131353 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112164 CK796080.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112164 AK185980.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112164 BC016133.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112164 BC029056.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112164 NM_144875.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141994 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00704620 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00704619 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00704617 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00100526 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00100523 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00704625 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00704629 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00100525 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20618102 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20618103 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20618104 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20618101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20618152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00704627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00335945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00704620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00635437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00095049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00620667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00085192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01117105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00620668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00095048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00704617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00066274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00704625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00704619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01117107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00100524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00100528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00620670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00620669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00335942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20618127 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10349731 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349729 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349727 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349732 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349730 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349728 - -
          Affymetrix Mouse Gene 1.0 ST Array 10349726 - -
          Affymetrix HT MG-430 PM Array Plate 1451362_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10349724 - -
          Mouse Genome 430 2.0 Array 1451362_at - -
          Mouse Genome 430 2.0 Array 1451362_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226422_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226422_at - -
          Affymetrix Mouse Clariom S Array PSR0100009567.mm.1 TC0100001177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009567.mm.1 NM_144875.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009567.mm.1 NM_144875.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009568.mm.1 TC0100001177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009568.mm.1 NM_144875.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009568.mm.1 NM_144875.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009569.mm.1 TC0100001177.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100009569.mm.1 NM_144875.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100009569.mm.1 NM_144875.2 Refseq
          Mouse Expression Set 430 1451362_PM_at - -
          Mouse Expression Set 430 1451362_at - -
          Mouse Genome 430A 2.0 Array 1451362_at - -
          Mouse Genome 430A 2.0 Array 1451362_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451362_at - -
          Affymetrix HT Mouse Genome 430A Array 1451362_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 647923 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 430450 - -
          Illumina mouse-6 v1.0 expression beadchip 430450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000430450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2681186 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2681186 - -
          mouse-6 v1.1 (Illumina) scl17491.6.1_47-S - -
          Illumina Sentrix mouse-6.v1 bead array 430450 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl17491.6.1_47-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 430450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2681186 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2681186 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl17491.6.1_47-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0