Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01290643_m1
          See other GM28856 GE Assays ›
          Gene Symbol:
          Gm28856
          Gene Name:
          predicted gene 28856
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 130765908 - 130781061 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01290643_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm28856
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK087224.1 - 511 61
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB664370.1 - 3 - 4 505 Chr. 1 - 130766388 - 130782240 [+] on Build GRCm38 61

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream expressed sequence AA986860 AA986860 E030025M07,
          Sarg
          -36.3kb Sense Chr. 1 - 130731976 - 130744622 on NCBI Build GRCm38
          Downstream Fc receptor,
          IgA,
          IgM,
          high affinity
          Fcamr 3.0kb Sense Chr. 1 - 130785260 - 130819316 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2281818 AK087224.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799337 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 419893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869016 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108162 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447486 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2281818 AK087224.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04262614 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04262597 - -
          Whole Mouse Genome G4122A A_51_P107633 AK087224.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125782 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04262614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20611692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20611691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20611682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00151411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00151415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00151414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20611680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00151417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00637241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20611686 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 305463 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0