Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291619_m1
          See other LYRM7OS GE Assays ›
          Gene Symbol:
          Lyrm7os
          Gene Name:
          LYR motif containing 7, opposite strand
          Gene Aliases:
          Gm12228, OTTMUSG00000005674
          Chromosome Location:
          Chr.11: 54819919 - 54824967 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291619_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lyrm7os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040284.1 2 - 3 429 121
          GenBank mRNA AK087691.1 - 2 - 3 430 121
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB553409.2 - 1 - 2 184 Chr. 11 - 54821328 - 54824967 [+] on Build GRCm38 121
          GenBankmRNA BB608639.2 - 2 - 3 428 Chr. 11 - 54819920 - 54824704 [+] on Build GRCm38 121

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream CDC42 small effector 2 Cdc42se2 2810404F18Rik,
          AA408783,
          AA536669,
          SPEC2
          -33.5kb Anti-sense Chr. 11 - 54717415 - 54787703 on NCBI Build GRCm38
          Downstream LYR motif containing 7 Lyrm7 1700024C24Rik,
          9330147L21Rik
          1.9kb Anti-sense Chr. 11 - 54826866 - 54860907 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000635.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192983 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143227 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07348816 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09528044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09528041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09265609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09157376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09528037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27812270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09447755 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100005138.mm.1 TC1100000635.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100005138.mm.1 NR_040284.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005138.mm.1 NR_040284.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 882407 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix