Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291698_m1
          See other 1700008C04RIK GE Assays ›
          Gene Symbol:
          1700008C04Rik
          Gene Name:
          RIKEN cDNA 1700008C04 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 51005156 - 51062543 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291698_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700008C04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK005755.1 - 4 - 5 561 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          40421
          Gm40421 -48.8kb Anti-sense Chr. 12 - 50985361 - 51003783 on NCBI Build GRCm38
          Downstream G2/M-phase specific E3 ubiquitin ligase G2e3 6030408C04Rik,
          9030416F18,
          AW046379,
          D930034K21Rik,
          mKIAA1333
          285.7kb Sense Chr. 12 - 51348230 - 51376986 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478962 AK013312.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478962 AK076122.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478962 BC008247.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478962 BC139829.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478962 CT010286.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478962 NM_007630.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478962 X66032.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647713 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757981 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17660 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07617571 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10013948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10125725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10137320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10291774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10144350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10408064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10266809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09930358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10420129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09844142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10173628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09987297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10173626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09839430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10013954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09769978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09777850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10081137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09747864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10144355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09839566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10120037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10189831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28428015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09955970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09769980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10144351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28427965 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100130292 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7f58f9545f-46b6w:80/100.66.130.62:80.
          git-commit: c36c52929dfaa9f2956040e6abd275853d6e1b95
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline